Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281437.1 | NP_001268366.1 | 7 | 1008 | ||
NM_001281438.1 | NP_001268367.1 | 7 | 1012 | |||
NM_007002.3 | NP_008933.2 | 8 | 1129 | |||
NM_175573.2 | NP_783163.1 | 8 | 1125 | |||
XM_005260257.1 | XP_005260314.1 | 9 | 1216 | |||
XM_011528503.1 | XP_011526805.1 | 8 | 1964 | |||
XM_017027602.1 | XP_016883091.1 | 8 | 1985 | |||
XM_017027603.1 | XP_016883092.1 | 9 | 1237 | |||
XM_017027604.1 | XP_016883093.1 | 8 | 1981 | |||
XM_017027605.1 | XP_016883094.1 | 8 | 1985 | |||
XM_017027606.1 | XP_016883095.1 | 7 | 1868 | |||
XM_017027607.1 | XP_016883096.1 | 7 | 1864 | |||
XM_017027608.1 | XP_016883097.1 | 6 | 1654 | |||
XM_017027609.1 | XP_016883098.1 | 6 | 1633 | |||
XM_017027610.1 | XP_016883099.1 | 6 | 1633 | |||
GenBank mRNA | AK054985.1 | - | 1926 | |||
AK307459.1 | - | 1059 | ||||
BC010733.1 | - | 5 | 641 | |||
BC017245.2 | - | 8 | 1025 | 4890786 | ||
CR749213.1 | - | 2 | 4541 | |||
D64154.1 | - | 8 | 1020 |