Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039141.2 | NP_001034230.1 | 14, 15 | 5851 | ||
NM_007032.5 | NP_008963.3 | 4, 5 | 445 | |||
NM_138632.2 | NP_619538.2 | 4, 5 | 445 | |||
GenBank mRNA | AF281030.1 | - | 4, 5 | 429 | ||
BC003618.2 | - | 4, 5 | 367 | |||
BC022200.1 | - | 4, 5 | 439 | |||
BQ880866.1 | - | 445 | ||||
DQ228003.1 | - | 12, 13 | 5514 | |||
DQ228005.1 | - | 13, 14 | 5716 |