Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPUL1 SIRNA Assays ›
Gene Symbol:
HNRNPUL1
Gene Name:
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases:
E1B-AP5, E1BAP5, HNRPUL1
Chromosome Location:
Chr.19: 41262476 - 41307783 on Build GRCh38
UniGene ID:
Hs.155218
Species:
Human
siRNA ID
HSS145968
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPUL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301016.2
NP_001287945.1
15
2521
NM_001321208.1
NP_001308137.1
15
2561
NM_001321211.1
NP_001308140.1
15
2345
NM_007040.5
NP_008971.2
15
2852
NM_144732.4
NP_653333.1
15
2334
XM_005258459.3
XP_005258516.1
15
2685
XM_005258461.4
XP_005258518.1
15
2515
XM_005258463.3
XP_005258520.1
15
2506
XM_005258464.3
XP_005258521.1
15
2321
XM_005258465.4
XP_005258522.1
15
2532
XM_011526392.2
XP_011524694.1
14
2463
XM_011526393.2
XP_011524695.1
15
2607
XM_011526394.2
XP_011524696.1
15
2362
XM_011526395.2
XP_011524697.1
15
2571
GenBank mRNA
AA479761.1
-
2
149
AJ007509.1
-
15
2695
AK001229.1
-
6
1118
AK021455.1
-
4
1137
AK022863.1
-
15
2654
AK023006.1
-
15
2291
AK127057.1
-
14
2342
AK225850.1
-
15
2344
AK290729.1
-
15
2316
AK291315.1
-
16
2601
AK291762.1
-
15
2654
AK297127.1
-
15
2449
AK308156.1
-
16
2364
AL050146.1
-
8
1460
BC002564.2
-
15
2592
3139049
BC004242.1
-
3
496
BC009988.2
-
15
2498
4127873
BC014232.2
-
15
2473
4558792
BC027713.2
-
15
2419
4123077
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
response to virus
RNA binding
protein binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline