Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003790.3 | NP_001003790.1 | 4 | 545 | ||
NM_001003791.2 | NP_001003791.1 | 4 | 316 | |||
NM_007175.6 | NP_009106.1 | 4 | 316 | |||
XM_005273392.2 | XP_005273449.1 | 4 | 476 | |||
XM_017013000.1 | XP_016868489.1 | 4 | 314 | |||
GenBank mRNA | AK291394.1 | - | 4 | 269 | ||
AK297279.1 | - | 4 | 269 | |||
AL442077.1 | - | 303 | ||||
AY358108.1 | - | 4 | 260 | |||
AY358851.1 | - | 4 | 250 | |||
BC005950.1 | - | 4 | 239 | |||
BC048308.1 | - | 4 | 539 | |||
BC050611.1 | - | 4 | 270 | |||
BC067765.1 | - | 4 | 543 | 5296776 | ||
BI560439.1 | - | 4 | 549 | |||
BP353279.1 | - | 4 | 316 |