Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007178.3 | NP_009109.3 | 9, 10 | 1351 | ||
XM_005253294.1 | XP_005253351.1 | 8, 9 | 1213 | |||
GenBank mRNA | AB024327.1 | - | 9, 10 | 1297 | ||
AF161496.1 | - | 9, 10 | 1298 | |||
AJ010025.1 | - | 9, 10 | 1144 | |||
AK297942.1 | - | 10, 11 | 1350 | |||
AK312295.1 | - | 9, 10 | 1292 | |||
AK315891.1 | - | 8, 9 | 1178 | |||
AL136691.1 | - | 9, 10 | 1293 | |||
AY049776.1 | - | 9, 10 | 1044 | |||
BC000162.2 | - | 9, 10 | 1142 | 2900954 | ||
BC062306.1 | - | 9, 10 | 1306 | 3349549 | ||
BT020044.1 | - | 9, 10 | 997 |
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641097.1 | - | 5 | 2288 |