Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007178.3 | NP_009109.3 | 10 | 1418 | ||
GenBank mRNA | AB024327.1 | - | 10 | 1364 | ||
AF161496.1 | - | 10 | 1365 | |||
AJ010025.1 | - | 10 | 1211 | |||
AK297942.1 | - | 11 | 1417 | |||
AK315891.1 | - | 9 | 1245 | |||
AL136691.1 | - | 10 | 1360 | |||
BC000162.2 | - | 10 | 1209 | 2900954 | ||
BC062306.1 | - | 10 | 1373 | 3349549 |