Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007178.3 | NP_009109.3 | 1 | 399 | ||
XM_005253294.1 | XP_005253351.1 | 1 | 397 | |||
GenBank mRNA | AB024327.1 | - | 1 | 345 | ||
AF161496.1 | - | 1 | 346 | |||
AJ010025.1 | - | 1 | 192 | |||
AK297942.1 | - | 1 | 359 | |||
AK312295.1 | - | 1 | 340 | |||
AK315891.1 | - | 1 | 362 | |||
AL136691.1 | - | 1 | 341 | |||
AY049776.1 | - | 1 | 92 | |||
BC000162.2 | - | 1 | 190 | 2900954 | ||
BC062306.1 | - | 1 | 354 | 3349549 | ||
BT020044.1 | - | 1 | 45 | |||
BT020045.1 | - | 1 | 45 | |||
DA666505.1 | - | 1 | 399 | |||
U56430.1 | - | 1 | 98 |