Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007213.2 | NP_009144.1 | 3 | 532 | ||
GenBank mRNA | AJ005896.1 | - | 3 | 490 | ||
AK023433.1 | - | 3 | 489 | |||
AK315374.1 | - | 3 | 489 | |||
BC021213.2 | - | 3 | 476 | 4297902 | ||
CR457023.1 | - | 3 | 474 |