Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281425.1 | NP_001268354.1 | 3 | 273 | ||
NM_001281426.1 | NP_001268355.1 | 3 | 273 | |||
NM_001281427.1 | NP_001268356.1 | 3 | 273 | |||
NM_007234.4 | NP_009165.1 | 3 | 273 | |||
NM_024348.3 | NP_077324.1 | 3 | 273 | |||
GenBank mRNA | AF082513.1 | - | 3 | 230 | ||
AK091661.1 | - | 3 | 268 | |||
AK314730.1 | - | 3 | 241 | |||
AW953496.1 | - | 3 | 218 | |||
BC000319.2 | - | 3 | 224 | 2821720 | ||
BC003004.1 | - | 3 | 224 | 2821720 | ||
BC107697.1 | - | 3 | 242 | 6454606 | ||
BG715167.1 | - | 3 | 254 | |||
BI596981.1 | - | 3 | 226 | |||
BI837796.1 | - | 3 | 232 | |||
CN257020.1 | - | 1 | 25 | |||
CR541889.1 | - | 3 | 214 | |||
DA212054.1 | - | 3 | 279 |