Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007238.4 | NP_009169.3 | 4 | 516 | ||
NM_183397.2 | NP_899634.1 | 3 | 317 | |||
GenBank mRNA | AF072864.1 | - | 4 | 438 | ||
AK023879.1 | - | 4 | 434 | |||
AK297018.1 | - | 3 | 306 | |||
AK301532.1 | - | 4 | 516 | |||
AK316589.1 | - | 4 | 496 | |||
AL136648.1 | - | 4 | 487 | |||
BC001147.1 | - | 4 | 417 | 3533572 | ||
BQ278083.1 | - | 3 | 285 | |||
CR533471.1 | - | 4 | 393 |