Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007282.4 | NP_009213.1 | 11 | 1905 | ||
NM_183381.2 | NP_899237.1 | 10 | 1803 | |||
XM_005247092.3 | XP_005247149.1 | 10 | 1762 | |||
XM_011512373.2 | XP_011510675.1 | 12 | 1910 | |||
XM_011512374.2 | XP_011510676.1 | 11 | 1808 | |||
XM_011512376.2 | XP_011510678.1 | 10 | 1213 | |||
XM_017005654.1 | XP_016861143.1 | 11 | 1984 | |||
XM_017005655.1 | XP_016861144.1 | 10 | 1885 | |||
XM_017005656.1 | XP_016861145.1 | 11 | 1864 | |||
XM_017005657.1 | XP_016861146.1 | 11 | 1997 | |||
XM_017005658.1 | XP_016861147.1 | 10 | 1895 | |||
XM_017005659.1 | XP_016861148.1 | 9 | 1632 | |||
XM_017005660.1 | XP_016861149.1 | 9 | 1622 | |||
XM_017005661.1 | XP_016861150.1 | 4 | 987 | |||
GenBank mRNA | AF037204.1 | - | 10 | 1272 | ||
AF070558.1 | - | 10 | 1263 | |||
AK090638.1 | - | 10 | 1783 | |||
AK090771.1 | - | 7 | 892 | |||
AK313304.1 | - | 10 | 1287 | |||
BC009781.1 | - | 10 | 1280 | 3683407 | ||
BC009803.1 | - | 11 | 1383 | 4274992 | ||
CR456804.1 | - | 9 | 1121 |