Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUF60 SIRNA Assays ›
Gene Symbol:
PUF60
Gene Name:
poly(U) binding splicing factor 60
Gene Aliases:
FIR, RoBPI, SIAHBP1, VRJS
Chromosome Location:
Chr.8: 143816344 - 143829386 on Build GRCh38
UniGene ID:
Hs.521924
Species:
Human
siRNA ID
HSS146066
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUF60
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136033.2
NP_001129505.1
12
1527
NM_001271096.1
NP_001258025.1
11
1454
NM_001271097.1
NP_001258026.1
10
1370
NM_001271098.1
NP_001258027.1
12
1505
NM_001271099.1
NP_001258028.1
11
1421
NM_001271100.1
NP_001258029.1
11
1476
NM_014281.4
NP_055096.2
11
1457
NM_078480.2
NP_510965.1
12
1508
XM_011516929.2
XP_011515231.1
13
1707
XM_011516930.2
XP_011515232.1
12
1558
XM_017013234.1
XP_016868723.1
12
3040
XM_017013235.1
XP_016868724.1
12
1610
XM_017013236.1
XP_016868725.1
11
2989
XM_017013237.1
XP_016868726.1
13
1769
XM_017013238.1
XP_016868727.1
13
1569
XM_017013239.1
XP_016868728.1
12
1535
XM_017013240.1
XP_016868729.1
11
1484
GenBank mRNA
AF114818.1
-
12
1489
AF190744.1
-
12
1417
AF217197.2
-
11
1384
AK292373.1
-
12
1489
AW249619.1
-
1
437
BC008875.2
-
11
1423
4138778
BC009734.1
-
11
1406
3877563
BC011265.1
-
12
1444
4155549
BC011979.1
-
10
1323
3863660
BC035435.2
-
11
1408
CR457143.1
-
12
1425
U51586.1
-
10
1371
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
RNA splicing
cell-cell adhesion
nucleotide binding
DNA binding
protein binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0