Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014981.1 | NP_055796.1 | 28 | 3609 | ||
XM_011512559.2 | XP_011510861.1 | 29 | 3730 | |||
XM_017005922.1 | XP_016861411.1 | 19 | 2874 | |||
GenBank mRNA | AB023217.3 | - | 28 | 3609 | ||
AK126801.1 | - | 10 | 1544 |