Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 6 | 546 | ||
NM_001144832.2 | NP_001138304.1 | 6 | 521 | |||
NM_001297662.1 | NP_001284591.1 | 6 | 628 | |||
NM_001297663.1 | NP_001284592.1 | 6 | 523 | |||
NM_001297664.1 | NP_001284593.1 | 6 | 559 | |||
NM_001297665.1 | NP_001284594.1 | 6 | 651 | |||
XM_005270644.2 | XP_005270701.1 | 6 | 552 | |||
XM_006710471.2 | XP_006710534.1 | 6 | 649 | |||
XM_011541048.2 | XP_011539350.1 | 6 | 626 | |||
XM_011541049.2 | XP_011539351.1 | 6 | 765 | |||
XM_011541050.2 | XP_011539352.1 | 6 | 626 | |||
XM_011541051.2 | XP_011539353.1 | 6 | 659 | |||
XM_011541052.2 | XP_011539354.1 | 6 | 605 | |||
XM_011541053.2 | XP_011539355.1 | 6 | 643 | |||
XM_011541054.2 | XP_011539356.1 | 6 | 479 | |||
XM_017000715.1 | XP_016856204.1 | 6 | 526 | |||
GenBank mRNA | AB007921.1 | - | 6 | 674 | ||
AF007170.1 | - | 6 | 427 | |||
AK000220.1 | - | 6 | 626 | |||
AK298019.1 | - | 6 | 530 | |||
AK301587.1 | - | 6 | 519 | |||
AK311430.1 | - | 6 | 448 | |||
BC028374.2 | - | 6 | 593 | |||
BI761577.1 | - | 6 | 664 | |||
CX788693.1 | - | 6 | 461 |