Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 7 | 615 | ||
NM_001144832.2 | NP_001138304.1 | 7 | 590 | |||
NM_001297662.1 | NP_001284591.1 | 7 | 697 | |||
NM_001297663.1 | NP_001284592.1 | 7 | 592 | |||
NM_001297664.1 | NP_001284593.1 | 7 | 628 | |||
NM_001297665.1 | NP_001284594.1 | 7 | 720 | |||
XM_005270644.2 | XP_005270701.1 | 7 | 621 | |||
XM_006710471.2 | XP_006710534.1 | 7 | 718 | |||
XM_011541048.2 | XP_011539350.1 | 7 | 695 | |||
XM_011541049.2 | XP_011539351.1 | 7 | 834 | |||
XM_011541050.2 | XP_011539352.1 | 7 | 695 | |||
XM_011541051.2 | XP_011539353.1 | 7 | 728 | |||
XM_011541052.2 | XP_011539354.1 | 7 | 674 | |||
XM_011541053.2 | XP_011539355.1 | 7 | 712 | |||
XM_011541054.2 | XP_011539356.1 | 7 | 548 | |||
XM_017000715.1 | XP_016856204.1 | 7 | 595 | |||
GenBank mRNA | AB007921.1 | - | 7 | 743 | ||
AF007170.1 | - | 7 | 496 | |||
AK000220.1 | - | 7 | 695 | |||
AK298019.1 | - | 7 | 599 | |||
AK301587.1 | - | 7 | 588 | |||
AK311430.1 | - | 7 | 517 | |||
BC028374.2 | - | 7 | 662 | |||
BI761577.1 | - | 7 | 733 | |||
CX788693.1 | - | 7 | 530 |