Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162499.1 | NP_001155971.1 | 10 | 2860 | ||
NM_012298.2 | NP_036430.1 | 8 | 2581 | |||
XM_011533503.2 | XP_011531805.1 | 10 | 2878 | |||
XM_011533504.2 | XP_011531806.1 | 10 | 2974 | |||
GenBank mRNA | AB014567.1 | - | 8 | 2509 | ||
BC136592.1 | - | 10 | 2799 | 9020582 | ||
BX502783.1 | - | 1 | 495 | |||
BX648800.1 | - | 1 | 13 | |||
GU727621.1 | - | 10 | 2860 |