Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015048.1 | NP_055863.1 | 5 | 860 | ||
XM_005253858.4 | XP_005253915.1 | 6 | 1019 | |||
XM_006719296.3 | XP_006719359.1 | 6 | 935 | |||
GenBank mRNA | JF813787.1 | - | 5 | 854 |