Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015060.2 | NP_055875.1 | 14, 15 | 2014 | ||
XM_005249668.2 | XP_005249725.1 | 14, 15 | 1963 | |||
XM_005249670.2 | XP_005249727.1 | 14, 15 | 1669 | |||
XM_017011891.1 | XP_016867380.1 | 14, 15 | 1657 | |||
XM_017011892.1 | XP_016867381.1 | 14, 15 | 1667 | |||
GenBank mRNA | AK090619.1 | - | 14, 15 | 1876 |