Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080424.1 | NP_001073893.1 | 19 | 4706 | ||
XM_005256549.3 | XP_005256606.1 | 20 | 4858 | |||
XM_005256550.4 | XP_005256607.1 | 19 | 4700 | |||
XM_005256551.3 | XP_005256608.1 | 19 | 4739 | |||
XM_005256552.4 | XP_005256609.1 | 19 | 5712 | |||
XM_006721483.3 | XP_006721546.1 | 20 | 4803 | |||
XM_011523750.2 | XP_011522052.1 | 20 | 6063 | |||
XM_011523752.2 | XP_011522054.1 | 19 | 4703 | |||
XM_017024381.1 | XP_016879870.1 | 19 | 6149 | |||
GenBank mRNA | AB002344.2 | - | 19 | 4711 | ||
BC009994.1 | - | 9 | 1335 |