Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015131.2 | NP_055946.1 | 12 | 1559 | ||
GenBank mRNA | AK000836.1 | - | 11 | 1255 | ||
D26488.1 | - | 12 | 1532 | |||
D87716.1 | - | 10 | 1038 |