Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323553.1 | NP_001310482.1 | 10 | 1298 | ||
NM_001323554.1 | NP_001310483.1 | 10 | 2237 | |||
NM_001323555.1 | NP_001310484.1 | 10 | 2306 | |||
NM_001323556.1 | NP_001310485.1 | 10 | 1079 | |||
NM_001323557.1 | NP_001310486.1 | 10 | 1148 | |||
NM_001323558.1 | NP_001310487.1 | 10 | 1229 | |||
NM_015137.5 | NP_055952.2 | 10 | 1229 | |||
NR_136615.1 | 10 | 1298 | ||||
NR_136616.1 | 10 | 1229 | ||||
GenBank mRNA | AK294485.1 | - | 4 | 368 | ||
AK307433.1 | - | 10 | 1180 | |||
BC071611.1 | - | 10 | 1206 | |||
BC108667.1 | - | 10 | 1197 | |||
BC131486.1 | - | 6 | 564 | |||
BC152442.1 | - | 10 | 1197 | 8862567 | ||
CR749243.1 | - | 10 | 1039 | |||
D63477.1 | - | 10 | 1197 |