Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014607.3 | NP_055422.1 | 12 | 1582 | ||
GenBank mRNA | AK026971.1 | - | 12 | 1205 | ||
AK095402.1 | - | 8 | 970 | |||
AK292829.1 | - | 12 | 1275 | |||
AK304096.1 | - | 12 | 1292 | |||
BC035594.2 | - | 13 | 1414 | 5580880 | ||
BX647216.1 | - | 10 | 2272 | |||
BX647662.1 | - | 12 | 1374 | |||
D87684.1 | - | 12 | 1335 | |||
EF199840.1 | - | 12 | 1399 |