Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014616.2 | NP_055431.1 | 7 | 828 | ||
XM_005247241.2 | XP_005247298.1 | 7 | 827 | |||
XM_005247243.2 | XP_005247300.1 | 5 | 419 | |||
XM_011512593.2 | XP_011510895.1 | 7 | 827 | |||
XM_011512594.2 | XP_011510896.1 | 7 | 827 | |||
XM_011512597.2 | XP_011510899.1 | 5 | 394 | |||
XM_017006008.1 | XP_016861497.1 | 5 | 396 | |||
XM_017006009.1 | XP_016861498.1 | 6 | 522 | |||
GenBank mRNA | AF156548.1 | - | 7 | 737 | ||
BX489704.1 | - | 3 | 243 |