Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015164.2 | NP_055979.2 | 13 | 2258 | ||
XM_005245790.3 | XP_005245847.1 | 12 | 2393 | |||
XM_005245791.4 | XP_005245848.1 | 8 | 1526 | |||
XM_017000757.1 | XP_016856246.1 | 13 | 2165 | |||
XM_017000758.1 | XP_016856247.1 | 12 | 2105 | |||
GenBank mRNA | AB020649.1 | - | 13 | 2034 | ||
BC008002.1 | - | 5 | 295 | |||
BC016488.1 | - | 5 | 940 | |||
BC030545.1 | - | 4 | 238 | |||
BC040441.1 | - | 13 | 2261 | |||
BC042103.1 | - | 5 | 588 | |||
BC068599.1 | - | 5 | 613 |