Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015168.1 | NP_055983.1 | 15 | 3461 | ||
XM_005258676.3 | XP_005258733.1 | 14 | 4736 | |||
XM_005258677.4 | XP_005258734.1 | 15 | 3587 | |||
XM_005258678.2 | XP_005258735.1 | 14 | 3344 | |||
XM_006723113.3 | XP_006723176.1 | 15 | 5151 | |||
XM_011526668.2 | XP_011524970.1 | 15 | 3363 | |||
XM_011526669.2 | XP_011524971.1 | 14 | 3491 | |||
XM_011526670.2 | XP_011524972.1 | 14 | 4087 | |||
XM_017026529.1 | XP_016882018.1 | 14 | 4543 | |||
XM_017026530.1 | XP_016882019.1 | 14 | 4899 | |||
XM_017026531.1 | XP_016882020.1 | 13 | 3242 | |||
GenBank mRNA | AB028987.2 | - | 15 | 3461 | ||
AK125279.1 | - | 7 | 1273 | |||
AL050155.1 | - | 1 | 449 |