Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE2 SIRNA Assays ›
Gene Symbol:
SYNE2
Gene Name:
spectrin repeat containing nuclear envelope protein 2
Gene Aliases:
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location:
Chr.14: 63761904 - 64226449 on Build GRCh38
UniGene ID:
Hs.745014
Species:
Human
siRNA ID
HSS146270
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015180.4
NP_055995.4
110
20074
NM_182910.2
NP_878914.1
4
805
NM_182913.2
NP_878917.1
6
910
NM_182914.2
NP_878918.2
111
20143
XM_005267454.1
XP_005267511.1
111
20143
XM_005267456.1
XP_005267513.1
111
20143
XM_005267457.1
XP_005267514.1
111
20143
XM_005267458.1
XP_005267515.1
110
20074
XM_005267459.1
XP_005267516.1
110
20074
XM_011536574.1
XP_011534876.1
111
20185
XM_011536575.2
XP_011534877.1
111
20162
XM_011536576.2
XP_011534878.1
112
20342
XM_011536577.2
XP_011534879.1
111
20109
XM_011536578.1
XP_011534880.1
111
20185
XM_011536579.1
XP_011534881.1
111
20185
XM_011536580.1
XP_011534882.1
111
20185
XM_011536581.1
XP_011534883.1
110
20116
XM_011536582.1
XP_011534884.1
110
20068
XM_017021101.1
XP_016876590.1
111
20187
XM_017021102.1
XP_016876591.1
110
20230
XM_017021103.1
XP_016876592.1
13
2222
XM_017021104.1
XP_016876593.1
13
2222
GenBank mRNA
AB023228.3
-
62
9815
AF357236.1
-
110
19613
AF435011.1
-
110
20056
AF495911.1
-
110
20041
AK074055.1
-
22
3541
AK090430.1
-
28
4450
AK297874.1
-
6
871
AK298465.1
-
5
585
AL080133.1
-
16
2587
AL117404.1
-
4
1327
AY061757.1
-
11
1678
AY061758.1
-
6
907
AY061759.1
-
58
9377
AY184204.1
-
4
1327
AY184205.1
-
9
1559
BX538095.1
-
19
3008
BX640974.1
-
12
1889
CR749324.1
-
31
5006
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nuclear envelope organization
nuclear migration
establishment or maintenance of cell polarity
fibroblast migration
positive regulation of cell migration
nuclear migration along microfilament
protein localization to nucleus
centrosome localization
cytoskeletal anchoring at nuclear membrane
actin binding
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0