Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015188.1 | NP_056003.1 | 4 | 1458 | ||
XM_006717732.3 | XP_006717795.2 | 4 | 1458 | |||
XM_006717733.2 | XP_006717796.1 | 5 | 418 | |||
XM_011539558.2 | XP_011537860.1 | 4 | 469 | |||
XM_017015996.1 | XP_016871485.1 | 4 | 1458 | |||
XM_017015997.1 | XP_016871486.1 | 4 | 1458 | |||
XM_017015998.1 | XP_016871487.1 | 3 | 245 | |||
XM_017015999.1 | XP_016871488.1 | 3 | 461 | |||
GenBank mRNA | AB011180.1 | - | 4 | 1274 | ||
AB449898.1 | - | 4 | 1255 | |||
AL049956.1 | - | 3 | 206 |