Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015207.1 | NP_056022.1 | 7 | 1029 | ||
XM_005245792.3 | XP_005245849.1 | 7 | 1688 | |||
XM_005245794.3 | XP_005245851.1 | 6 | 664 | |||
GenBank mRNA | AB007928.3 | - | 7 | 1006 | ||
BC119636.2 | - | 3 | 247 |