Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015279.1 | NP_056094.1 | 8, 9 | 1340 | ||
XM_011538078.2 | XP_011536380.1 | 8, 9 | 1282 | |||
XM_017019083.1 | XP_016874572.1 | 8, 9 | 1282 | |||
XM_017019084.1 | XP_016874573.1 | 9, 10 | 941 | |||
XM_017019085.1 | XP_016874574.1 | 10, 11 | 1221 | |||
XM_017019086.1 | XP_016874575.1 | 10, 11 | 1226 | |||
XM_017019087.1 | XP_016874576.1 | 8, 9 | 1092 | |||
GenBank mRNA | AB023201.1 | - | 9, 10 | 935 | ||
AB449914.1 | - | 8, 9 | 1032 | |||
AK055404.1 | - | 8, 9 | 1282 | |||
AK299508.1 | - | 7, 8 | 1119 |