Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 37 | 4787 | ||
XM_011525642.1 | XP_011523944.1 | 37 | 4787 | |||
XM_011525643.2 | XP_011523945.1 | 37 | 4787 | |||
XM_011525644.2 | XP_011523946.1 | 36 | 4695 | |||
XM_017025684.1 | XP_016881173.1 | 38 | 4965 | |||
GenBank mRNA | AB014550.1 | - | 11 | 1129 | ||
AK126324.1 | - | 22 | 2418 | |||
AL080138.1 | - | 8 | 767 | |||
CR627458.1 | - | 26 | 3011 |