Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304333.2 | NP_001291262.1 | 2 | 546 | ||
NM_015297.2 | NP_056112.1 | 2 | 542 | |||
NR_136307.1 | 2 | 546 | ||||
NR_136308.1 | 2 | 542 | ||||
XM_011517827.1 | XP_011516129.1 | 3 | 531 | |||
XM_011517828.2 | XP_011516130.1 | 2 | 635 | |||
XM_011517829.1 | XP_011516131.1 | 2 | 793 | |||
XM_017014553.1 | XP_016870042.1 | 4 | 825 | |||
XM_017014554.1 | XP_016870043.1 | 4 | 605 | |||
XM_017014555.1 | XP_016870044.1 | 4 | 577 | |||
XM_017014556.1 | XP_016870045.1 | 4 | 1109 | |||
GenBank mRNA | AB028968.1 | - | 2 | 413 | ||
AK294342.1 | - | 2 | 546 |