Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 35 | 3136 | ||
NM_015303.3 | NP_056118.2 | 34 | 3130 | |||
XM_005247251.3 | XP_005247308.1 | 35 | 3404 | |||
XM_005247253.4 | XP_005247310.1 | 35 | 3142 | |||
XM_006713556.3 | XP_006713619.1 | 27 | 2396 | |||
XM_011512599.2 | XP_011510901.1 | 22 | 1902 | |||
XM_011512600.2 | XP_011510902.1 | 28 | 2423 | |||
XM_011512601.2 | XP_011510903.1 | 28 | 2603 | |||
XM_017006040.1 | XP_016861529.1 | 33 | 2977 | |||
GenBank mRNA | AB018347.1 | - | 27 | 2315 | ||
AK022945.1 | - | 13 | 1145 | |||
AK056661.1 | - | 35 | 3108 | |||
AK292423.1 | - | 34 | 3102 | |||
AK309244.1 | - | 14 | 1372 | |||
AL833838.1 | - | 32 | 2669 | |||
BC140768.1 | - | 35 | 3072 | 9021636 | ||
BI914013.1 | - | 1 | 59 | |||
BX647915.1 | - | 32 | 2821 |