Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 42 | 3734 | ||
NM_015303.3 | NP_056118.2 | 41 | 3728 | |||
XM_005247251.3 | XP_005247308.1 | 42 | 4002 | |||
XM_006713556.3 | XP_006713619.1 | 34 | 2994 | |||
XM_011512599.2 | XP_011510901.1 | 29 | 2500 | |||
XM_011512600.2 | XP_011510902.1 | 35 | 3021 | |||
XM_011512601.2 | XP_011510903.1 | 35 | 3201 | |||
XM_017006040.1 | XP_016861529.1 | 40 | 3575 | |||
GenBank mRNA | AB018347.1 | - | 34 | 2913 | ||
AK022945.1 | - | 20 | 1743 | |||
AK292423.1 | - | 41 | 3700 | |||
AK309244.1 | - | 21 | 1970 | |||
AL833838.1 | - | 39 | 3267 | |||
BC140768.1 | - | 42 | 3670 | 9021636 | ||
BI914013.1 | - | 8 | 661 | |||
BX647915.1 | - | 39 | 3419 |