Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173408.1 | NP_001166879.1 | 2 | 1384 | ||
NM_001173431.1 | NP_001166902.1 | 2 | 1384 | |||
NM_015311.2 | NP_056126.1 | 2 | 1384 | |||
XM_005246424.4 | XP_005246481.1 | 2 | 1408 | |||
XM_005246427.4 | XP_005246484.1 | 2 | 1399 | |||
XM_011510857.2 | XP_011509159.1 | 2 | 1411 | |||
XM_011510863.2 | XP_011509165.1 | 2 | 1403 | |||
XM_011510864.2 | XP_011509166.1 | 2 | 1403 | |||
XM_011510865.2 | XP_011509167.1 | 2 | 1403 | |||
XM_011510866.2 | XP_011509168.1 | 2 | 1402 | |||
XM_017003696.1 | XP_016859185.1 | 2 | 1410 | |||
XM_017003697.1 | XP_016859186.1 | 2 | 1410 | |||
XM_017003698.1 | XP_016859187.1 | 2 | 1409 | |||
XM_017003699.1 | XP_016859188.1 | 2 | 1410 | |||
XM_017003700.1 | XP_016859189.1 | 2 | 1408 | |||
GenBank mRNA | AB014557.2 | - | 2 | 579 | ||
BC007201.1 | - | 2 | 1129 | |||
EF063637.1 | - | 2 | 1127 | |||
EF063638.1 | - | 2 | 1127 |