Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 15 | 2282 | ||
NM_001300952.1 | NP_001287881.1 | 15 | 2282 | |||
NM_015326.4 | NP_056141.2 | 15 | 2285 | |||
XM_005277510.2 | XP_005277567.1 | 16 | 2472 | |||
XM_005277511.2 | XP_005277568.1 | 16 | 2469 | |||
XM_005277514.1 | XP_005277571.1 | 16 | 2472 | |||
XM_005277515.1 | XP_005277572.1 | 16 | 2469 | |||
XM_011509354.1 | XP_011507656.1 | 16 | 2472 | |||
XM_011509355.1 | XP_011507657.1 | 16 | 2469 | |||
XM_011509356.2 | XP_011507658.1 | 16 | 2472 | |||
XM_011509359.1 | XP_011507661.1 | 16 | 2472 | |||
XM_017000838.1 | XP_016856327.1 | 14 | 2116 | |||
XM_017000839.1 | XP_016856328.1 | 14 | 2146 | |||
XM_017000840.1 | XP_016856329.1 | 16 | 1761 | |||
XM_017000841.1 | XP_016856330.1 | 16 | 2469 | |||
GenBank mRNA | AB007925.1 | - | 15 | 1815 | ||
AK293335.1 | - | 12 | 1321 | |||
AK295845.1 | - | 15 | 2038 | |||
BC041635.1 | - | 7 | 586 | |||
BC132872.1 | - | 15 | 1813 | 40146894 | ||
BC132874.1 | - | 15 | 1813 | 40146896 | ||
BC144343.1 | - | 15 | 1810 | 9052863 |