Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080398.1 | NP_001073867.1 | 19 | 2265 | ||
XM_005251850.3 | XP_005251907.1 | 17 | 1768 | |||
XM_005251851.4 | XP_005251908.1 | 18 | 2147 | |||
XM_005251852.4 | XP_005251909.1 | 18 | 2099 | |||
XM_005251853.3 | XP_005251910.1 | 18 | 1881 | |||
XM_005251854.3 | XP_005251911.1 | 17 | 2050 | |||
XM_011518422.2 | XP_011516724.1 | 17 | 1768 | |||
XM_017014557.1 | XP_016870046.1 | 11 | 1087 | |||
GenBank mRNA | AB002366.1 | - | 7 | 539 | ||
AK025689.1 | - | 2 | 67 | |||
AK122712.1 | - | 17 | 1752 | |||
AK127247.1 | - | 13 | 1206 | |||
AK308313.1 | - | 18 | 1872 |