Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193267.2 | NP_001180196.1 | 3, 4 | 404 | ||
NM_012241.4 | NP_036373.1 | 3, 4 | 426 | |||
NM_031244.3 | NP_112534.1 | 3, 4 | 404 | |||
XM_005248968.4 | XP_005249025.1 | 3, 4 | 341 | |||
XM_017010620.1 | XP_016866109.1 | 2, 3 | 304 | |||
XM_017010621.1 | XP_016866110.1 | 3, 4 | 341 | |||
XM_017010624.1 | XP_016866113.1 | 2, 3 | 304 | |||
XM_017010625.1 | XP_016866114.1 | 2, 3 | 304 | |||
GenBank mRNA | AF083110.1 | - | 3, 4 | 387 | ||
AJ420496.1 | - | 3, 4 | 333 | |||
AK000355.1 | - | 3, 4 | 332 | |||
AK302467.1 | - | 3, 4 | 339 | |||
BC000126.1 | - | 3, 4 | 410 | 3352592 | ||
CB155518.1 | - | 3, 4 | 404 | |||
DC314190.1 | - | 3, 4 | 426 |