Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178074.1 | NP_001171545.1 | 6 | 755 | ||
NM_021150.3 | NP_066973.2 | 6 | 755 | |||
XM_005268754.4 | XP_005268811.1 | 6 | 675 | |||
XM_005268757.4 | XP_005268814.1 | 5 | 594 | |||
XM_011538093.2 | XP_011536395.1 | 6 | 728 | |||
XM_017019098.1 | XP_016874587.1 | 6 | 1024 | |||
XM_017019099.1 | XP_016874588.1 | 6 | 3683 | |||
XM_017019100.1 | XP_016874589.1 | 6 | 1024 | |||
XM_017019101.1 | XP_016874590.1 | 6 | 3683 | |||
XM_017019102.1 | XP_016874591.1 | 6 | 3683 | |||
GenBank mRNA | AK095519.1 | - | 6 | 755 | ||
AK295565.1 | - | 5 | 561 | |||
AK307576.1 | - | 6 | 593 | |||
BC029871.1 | - | 5 | 475 | |||
BC115393.1 | - | 6 | 583 | 40053971 | ||
BC115394.1 | - | 6 | 583 | 40053972 |