Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012205.2 | NP_036337.2 | 6, 7 | 561 | ||
XM_011532729.2 | XP_011531031.1 | 5, 6 | 2219 | |||
XM_011532730.2 | XP_011531032.1 | 7, 8 | 655 | |||
GenBank mRNA | BC029510.1 | - | 6, 7 | 544 | 5302724 | |
BI602406.1 | - | 6, 7 | 549 | |||
CR457063.1 | - | 6, 7 | 486 | |||
DT215851.1 | - | 6, 7 | 563 | |||
Z29481.1 | - | 6, 7 | 526 |