Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012404.2 | NP_036536.2 | 1 | 13 | ||
GenBank mRNA | BC069122.1 | - | 1 | 13 | 7216976 |
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC106719.2 | - | 1 | 13 | 40014106 |