Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080671.3 | NP_542402.3 | 2 | 859 | ||
GenBank mRNA | AK294423.1 | - | 2 | 859 | ||
AK313203.1 | - | 2 | 599 | |||
AY065987.1 | - | 2 | 573 | |||
BC014429.1 | - | 2 | 580 | 4548085 |