Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317996.1 | NP_001304925.1 | 12 | 983 | ||
NM_014337.3 | NP_055152.1 | 12 | 983 | |||
NM_148175.2 | NP_680480.1 | 12 | 983 | |||
NM_148176.2 | NP_680481.1 | 12 | 983 | |||
XM_005261448.3 | XP_005261505.1 | 12 | 983 | |||
XM_011530041.2 | XP_011528343.1 | 12 | 983 | |||
XM_011530042.2 | XP_011528344.1 | 12 | 983 | |||
XM_011530043.2 | XP_011528345.1 | 12 | 983 | |||
XM_011530044.2 | XP_011528346.1 | 12 | 983 | |||
XM_011530046.2 | XP_011528348.1 | 12 | 983 | |||
XM_011530047.2 | XP_011528349.1 | 12 | 983 | |||
XM_011530049.2 | XP_011528351.1 | 13 | 1033 | |||
XM_011530050.2 | XP_011528352.1 | 11 | 896 | |||
XM_011530051.2 | XP_011528353.1 | 12 | 923 | |||
XM_017028705.1 | XP_016884194.1 | 3 | 229 | |||
XM_017028706.1 | XP_016884195.1 | 3 | 229 | |||
GenBank mRNA | AK127259.1 | - | 14 | 1095 | ||
AK289545.1 | - | 12 | 945 | |||
AK299206.1 | - | 11 | 845 | |||
BC000022.2 | - | 12 | 945 | 3505936 | ||
BC028385.2 | - | 12 | 927 | 4816793 | ||
CR456548.1 | - | 12 | 899 | |||
U37219.1 | - | 12 | 959 | |||
U37220.1 | - | 3 | 207 | |||
U37221.1 | - | 3 | 242 |