Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012109.2 | NP_036241.1 | 4 | 612 | ||
GenBank mRNA | AF186264.1 | - | 4 | 582 | ||
AK309384.1 | - | 4 | 611 | |||
BC010446.1 | - | 4 | 553 | 4181442 | ||
BI752768.1 | - | 4 | 622 |