Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012094.4 | NP_036226.1 | 6 | 752 | ||
NM_181651.2 | NP_857634.1 | 5 | 620 | |||
NM_181652.2 | NP_857635.1 | 4 | 485 | |||
XM_005273882.4 | XP_005273939.2 | 5 | 588 | |||
GenBank mRNA | AA187737.1 | - | 1 | 157 | ||
AF110731.1 | - | 6 | 660 | |||
AF112212.1 | - | 6 | 651 | |||
AF124993.1 | - | 6 | 693 | |||
AF197952.1 | - | 6 | 733 | |||
AF231705.1 | - | 6 | 643 | |||
AF242525.1 | - | 6 | 659 | |||
AJ249483.1 | - | 5 | 468 | |||
BC110983.1 | - | 6 | 752 | 5166149 | ||
BC113723.1 | - | 6 | 658 | 8322775 | ||
BC113725.1 | - | 6 | 658 | 8322777 | ||
BC143849.1 | - | 5 | 526 | 9052365 | ||
BC171733.1 | - | 4 | 391 | 9052364 | ||
CR457203.1 | - | 6 | 624 | |||
FJ224298.1 | - | 5 | 468 |