Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014351.3 | NP_055166.1 | 4 | 608 | ||
XM_011530120.2 | XP_011528422.1 | 4 | 542 | |||
GenBank mRNA | AF115311.1 | - | 4 | 492 | ||
AF176342.1 | - | 4 | 512 | |||
AF188698.1 | - | 4 | 575 | |||
AF251263.1 | - | 4 | 521 | |||
AK295399.1 | - | 4 | 441 | |||
AK313048.1 | - | 4 | 533 | |||
AL590119.1 | - | 4 | 492 | |||
BC022459.1 | - | 4 | 501 | 4795827 | ||
BC028171.1 | - | 4 | 580 | 5243984 | ||
BC030665.1 | - | 4 | 636 | 5258865 | ||
CR456588.1 | - | 4 | 493 |