Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100819.2 | NP_001094289.1 | 5, 6 | 418 | ||
NM_001204094.1 | NP_001191023.1 | 6, 7 | 435 | |||
NM_015387.4 | NP_056202.2 | 6, 7 | 481 | |||
NM_199482.3 | NP_955776.1 | 6, 7 | 491 | |||
GenBank mRNA | AB015441.1 | - | 6, 7 | 440 | ||
AF093825.1 | - | 6, 7 | 456 | |||
AF151853.1 | - | 6, 7 | 456 | |||
AF250319.1 | - | 6, 7 | 466 | |||
AJ580638.1 | - | 6, 7 | 437 | |||
AK027043.1 | - | 6, 7 | 478 | |||
AK292938.1 | - | 6, 7 | 477 | |||
AK297294.1 | - | 352 | ||||
AK297514.1 | - | 5, 6 | 377 | |||
AK307497.1 | - | 5, 6 | 349 | |||
AL080070.1 | - | 6, 7 | 450 | |||
BC005237.1 | - | 6, 7 | 481 | 3996585 | ||
CR457371.1 | - | 6, 7 | 437 | |||
DC298610.1 | - | 6, 7 | 491 | |||
DC350915.1 | - | 5, 6 | 414 | |||
DC375649.1 | - | 6, 7 | 437 |
Gene Symbol: HSPE1-MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202485.1 | NP_001189414.1 | 7, 8 | 985 | ||
GenBank mRNA | CR990871.1 | - | 7, 8 | 569 |