Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015473.3 | NP_056288.2 | 29 | 4856 | ||
GenBank mRNA | AB037737.1 | - | 20 | 3304 | ||
AK000343.1 | - | 7 | 1142 | |||
AK023789.1 | - | 11 | 1863 | |||
BC045764.1 | - | 23 | 3869 | |||
BC063867.1 | - | 11 | 1880 | |||
CR749444.1 | - | 22 | 3554 |