Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042635.1 | NP_001036100.1 | 3 | 154 | ||
NM_015514.1 | NP_056329.1 | 3 | 154 | |||
GenBank mRNA | AK022215.1 | - | 3 | 136 | ||
AK291875.1 | - | 3 | 147 | |||
AL050003.1 | - | 3 | 142 | |||
BC030817.1 | - | 3 | 140 | 4768089 |