Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 10 | 1142 | ||
NR_046462.1 | 9 | 1105 | ||||
XM_005248480.2 | XP_005248537.1 | 10 | 1277 | |||
XM_011543306.2 | XP_011541608.1 | 10 | 1364 | |||
XM_011543307.2 | XP_011541609.1 | 10 | 1793 | |||
XM_017009332.1 | XP_016864821.1 | 8 | 2040 | |||
XM_017009333.1 | XP_016864822.1 | 8 | 1014 | |||
GenBank mRNA | AB020695.1 | - | 10 | 1094 | ||
AK022981.1 | - | 1 | 98 | |||
AK290004.1 | - | 10 | 1142 | |||
BX640902.1 | - | 9 | 923 | |||
BX647536.1 | - | 9 | 1107 | |||
CR936791.1 | - | 10 | 1144 |