Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145638.2 | NP_001139110.2 | 14 | 2109 | ||
NM_001145639.1 | NP_001139111.1 | 4 | 533 | |||
NM_001200003.1 | NP_001186932.1 | 4 | 657 | |||
XM_011518498.2 | XP_011516800.1 | 14 | 2000 | |||
XM_017014598.1 | XP_016870087.1 | 13 | 1908 | |||
XM_017014599.1 | XP_016870088.1 | 6 | 875 | |||
GenBank mRNA | AL117478.1 | - | 8 | 949 | ||
AL545250.3 | - | 4 | 657 | |||
AY173053.1 | - | 13 | 1820 | |||
BC048343.1 | - | 3 | 594 | |||
BI766024.1 | - | 4 | 564 | |||
BQ214873.1 | - | 1 | 53 |